Learn More
Chemglass Life Sciences Stabilizer Bars, Set of Three, for 1-3L Bioreactor Stands
SDP
Catalog No. 5000083198
Encompass
Bars attach to the legs of bioreactor stands with two adjustable hex head screws and add an additional measure of safety.For use with both jacketed and unjacketed bioreactor stands.Each bar has silicone pads on the ends for cushioning and added extra grip.Sold as a set of three. 1-3L stabilizer bars are 4” long5-7 stabilizer bars are 4” long15L stabilizer bars are 6” long
Catalog No. 50-000-83198 Supplier Chemglass Life Sciences Supplier No. CLS1380SBD3
Bars attach to the legs of bioreactor stands with two adjustable hex head screws and add an additional measure of safety.For use with both jacketed and unjacketed bioreactor stands.Each bar has silicone pads on the ends for cushioning and added extra grip.Sold as a set of three. 1-3L stabilizer bars are 4” long5-7 stabilizer bars are 4” long15L stabilizer bars are 6” long