Learn More
Electron Microscopy Sciences Water Bath Stabilizing Ring for 5-10 ml cylinders, 165 gm, 20 mm
SDP
Catalog No. 5028985
Encompass_Preferred
Vinyl coated lead rings which stabilize glassware that is immersed in water baths; reduces tilting and chemical spills.
Weight: 165 gm
I.D.: 20 mm
Fits: 5-10ml cylinders
Catalog No. 50-289-85 Supplier Electron Microscopy Sciences Supplier No. 64435
Vinyl coated lead rings which stabilize glassware that is immersed in water baths; reduces tilting and chemical spills.
Weight: 165 gm
I.D.: 20 mm
Fits: 5-10ml cylinders