Learn More
Electron Microscopy Sciences Water Bath Stabilizing Ring for 250-1000 ml flasks, 454 gm, 56 mm
SDP
Catalog No. 5028983
Encompass_Preferred
Vinyl coated lead rings which stabilize glassware that is immersed in water baths; reduces tilting and chemical spills.
Weight: 454 gm
I.D.: 56 mm
Fits: 250-1000ml flasks
Catalog No. 50-289-83 Supplier Electron Microscopy Sciences Supplier No. 64437
Vinyl coated lead rings which stabilize glassware that is immersed in water baths; reduces tilting and chemical spills.
Weight: 454 gm
I.D.: 56 mm
Fits: 250-1000ml flasks