Learn More
Electron Microscopy Sciences Water Bath Stabilizing Ring for 500-2000 ml flasks, 681 gm, 61 mm
SDP
Catalog No. 5028986
Encompass_Preferred
Vinyl coated lead rings which stabilize glassware that is immersed in water baths; reduces tilting and chemical spills.
Weight: 681 gm
I.D.: 61 mm
Fits: 500-2000ml flasks
Catalog No. 50-289-86 Supplier Electron Microscopy Sciences Supplier No. 64438
Add to Cart
Add to Cart
Vinyl coated lead rings which stabilize glassware that is immersed in water baths; reduces tilting and chemical spills.
Weight: 681 gm
I.D.: 61 mm
Fits: 500-2000ml flasks