Learn More
Electron Microscopy Sciences Water Bath Stabilizing Ring for 1000-4000 ml flasks, 908 gm, 74 mm
SDP
Catalog No. 5028984
Encompass_Preferred
Vinyl coated lead rings which stabilize glassware that is immersed in water baths; reduces tilting and chemical spills.
Weight: 908 gm
I.D.: 74 mm
Fits: 1000-4000ml flasks
Catalog No. 50-289-84 Supplier Electron Microscopy Sciences Supplier No. 64439
Vinyl coated lead rings which stabilize glassware that is immersed in water baths; reduces tilting and chemical spills.
Weight: 908 gm
I.D.: 74 mm
Fits: 1000-4000ml flasks