Learn More
Surmodics Inc StabilZyme Protein-Free Stabilizer, 50mL
SDP
Catalog No. NC1640788
Encompass_Preferred
StabilZyme HRP, StabilZyme Protein-Free and StabilZyme NOBLE stabilizers provide the assay developer a complete set of tools to stabilize different HRP conjugates for long term, in-solution stability.
Catalog No. NC1640788 Supplier Surmodics Inc Supplier No. SZPF0050
StabilZyme HRP, StabilZyme Protein-Free and StabilZyme NOBLE stabilizers provide the assay developer a complete set of tools to stabilize different HRP conjugates for long term, in-solution stability.