Learn More
Surmodics Inc StabilZyme™ SELECT Stabilizer, 50mL
SDP
Catalog No. NC0701324
Encompass_Preferred
The StabilZyme SELECT Stabilizer is a multi-purpose stabilizing solution designed to maintain the conformation of antibody/antigen conjugates, antibody coated particles and unmodified proteins that are commonly used in immunoassays. Adding the conjugate/protein directly into the SELECT Stabilizer allows for stable storage of the conjugate at a working concentration and eliminates the need for subsequent dilutions.
Catalog No. NC0701324 Supplier Surmodics Inc Supplier No. SZ030050
The StabilZyme SELECT Stabilizer is a multi-purpose stabilizing solution designed to maintain the conformation of antibody/antigen conjugates, antibody coated particles and unmodified proteins that are commonly used in immunoassays. Adding the conjugate/protein directly into the SELECT Stabilizer allows for stable storage of the conjugate at a working concentration and eliminates the need for subsequent dilutions.