Learn More
Hach Company Stablcal Turbidity Standards Calibration Set, 2100Q Portable Turbidimeter, 100 mL bottles
Catalog No. 501996970 Shop All Hach Company Products
Encompass
Stablcal Calibration Standards Kit, for 2100Q Portable Turbidimeter. USEPA accepted for calibration purposes. Stablcal Stabilized Formazin Turbidity Standards are true Formazin dilutions developed for use in any turbidimeter. Kit includes 100 mL bottles of 10, 20, 100 and 800 NTU standard.
Catalog No. 50-199-6970 Supplier Hach Company Supplier No. 2971210
Stablcal Calibration Standards Kit, for 2100Q Portable Turbidimeter. USEPA accepted for calibration purposes. Stablcal Stabilized Formazin Turbidity Standards are true Formazin dilutions developed for use in any turbidimeter. Kit includes 100 mL bottles of 10, 20, 100 and 800 NTU standard.