Learn More
Hach Company Stablcal Turbidity Standards Calibration Kit, 2100Q Portable Turbidimeter, 500 mL bottles
Catalog No. 501996968 Shop All Hach Company Products
Encompass
Stablcal Calibration Standards Kit, for 2100Q Portable Turbidimeter. USEPA accepted for calibration purposes. Stablcal Stabilized Formazin Turbidity Standards are true Formazin dilutions developed for use in any turbidimeter. Kit includes 500 mL bottles of 10, 20, 100 and 800 NTU standard.
Catalog No. 50-199-6968 Supplier Hach Company Supplier No. 2971200
Stablcal Calibration Standards Kit, for 2100Q Portable Turbidimeter. USEPA accepted for calibration purposes. Stablcal Stabilized Formazin Turbidity Standards are true Formazin dilutions developed for use in any turbidimeter. Kit includes 500 mL bottles of 10, 20, 100 and 800 NTU standard.