Learn More
Hach Company Stablcal Turbidity Standards Calibration Kit, 2100Q/QIS Portable Turbidimeter, Sealed Vials
Catalog No. 501996969 Shop All Hach Company Products
Encompass
Stablcal Calibration Standards Kit, for 2100Q Portable Turbidimeter.Stablcal Stabilized Formazin Turbidity Standards are true Formazin dilutions developed for use in any turbidimeter. Kit contains one each of 20, 100, and 800 NTU Stablcal vials for calibration and one 10 NTU Stablcal vial for verification.
Catalog No. 50-199-6969 Supplier Hach Company Supplier No. 2971205
Stablcal Calibration Standards Kit, for 2100Q Portable Turbidimeter.Stablcal Stabilized Formazin Turbidity Standards are true Formazin dilutions developed for use in any turbidimeter. Kit contains one each of 20, 100, and 800 NTU Stablcal vials for calibration and one 10 NTU Stablcal vial for verification.