Learn More
Hach Company Stablcal Turbidity Standards Calibration Kit, 0-4000 NTU, Sealed Vials
Catalog No. 501996500 Shop All Hach Company Products
Encompass
Stablcal Stabilized Formazin Turbidity Standards are true Formazin dilutions developed for use in any turbidimeter. Kit includes sealed vials of 0.1, 20, 200, 1000 and 4000 NTU standards.For use with the 2100N, 2100N IS, TL2300 and TL2310 IS turbidimeter.
Catalog No. 50-199-6500 Supplier Hach Company Supplier No. 2662105
Stablcal Stabilized Formazin Turbidity Standards are true Formazin dilutions developed for use in any turbidimeter. Kit includes sealed vials of 0.1, 20, 200, 1000 and 4000 NTU standards.For use with the 2100N, 2100N IS, TL2300 and TL2310 IS turbidimeter.