Learn More
Hach Company Stablcal Turbidity Standards Calibration Kit, 0 - 7500 NTU, Sealed Vials
Catalog No. 501996474 Shop All Hach Company Products
Encompass
Stablcal Standards Calibration Kit, for 2100AN, 2100AN IS, TL2350 and TL2360 IS Laboratory Turbidimeters. Stablcal Stabilized Formazin Turbidity Standards are true Formazin dilutions developed for use in any turbidimeter. Kit includes sealed vials of 0.1, 20, 200, 1000, 4000 NTU and 7500 NTU standard.
Catalog No. 50-199-6474 Supplier Hach Company Supplier No. 2659505
Stablcal Standards Calibration Kit, for 2100AN, 2100AN IS, TL2350 and TL2360 IS Laboratory Turbidimeters. Stablcal Stabilized Formazin Turbidity Standards are true Formazin dilutions developed for use in any turbidimeter. Kit includes sealed vials of 0.1, 20, 200, 1000, 4000 NTU and 7500 NTU standard.