Learn More
Hach Company Stablcal Turbidity Standards Calibration Kit, 2100A Turbidimeter, Sealed Vials
Catalog No. 501996468 Shop All Hach Company Products
Encompass
Stablcal Standards Calibration Kit, for 2100A Laboratory Turbidimeter. USEPA accepted for calibration purposes. Stablcal Stabilized Formazin Turbidity Standards are true Formazin dilutions developed for use in any turbidimeter. Kit includes Sealed Vials - 1.0, 10, 100 and 1000 NTU
Catalog No. 50-199-6468 Supplier Hach Company Supplier No. 2659105
Stablcal Standards Calibration Kit, for 2100A Laboratory Turbidimeter. USEPA accepted for calibration purposes. Stablcal Stabilized Formazin Turbidity Standards are true Formazin dilutions developed for use in any turbidimeter. Kit includes Sealed Vials - 1.0, 10, 100 and 1000 NTU