Learn More
Hach Company Stablcal Turbidity Standards Calibration Kit, 2100P Portable Turbidimeter, Sealed Vials
Catalog No. 501996471 Shop All Hach Company Products
Encompass
Stablcal Calibration Standards Kit, for 2100P Portable Turbidimeter. Stablcal Stabilized Formazin Turbidity Standards are true Formazin dilutions developed for use in any turbidimeter. Kit includes Sealed Vials of 0.1, 20, 100 and 800 NTU standard.
Catalog No. 50-199-6471 Supplier Hach Company Supplier No. 2659405
Stablcal Calibration Standards Kit, for 2100P Portable Turbidimeter. Stablcal Stabilized Formazin Turbidity Standards are true Formazin dilutions developed for use in any turbidimeter. Kit includes Sealed Vials of 0.1, 20, 100 and 800 NTU standard.