Learn More
Hach Company Stablcal Turbidity Standards Calibration Kit, 2100AN / AN IS Turbidimeter, 100 mL bottles
Catalog No. 501996475 Shop All Hach Company Products
Encompass
Stablcal Standards Calibration Kit, for 2100AN and 2100ANis Laboratory Turbidimeters. USEPA accepted for calibration purposes. Stablcal Stabilized Formazin Turbidity Standards are true Formazin dilutions developed for use in any turbidimeter. Kit includes 100 mL bottles of 0.1, 20, 200, 1000, 4000 NTU standard and a sealed vial of 7500 NTU standard.
Catalog No. 50-199-6475 Supplier Hach Company Supplier No. 2659510
Stablcal Standards Calibration Kit, for 2100AN and 2100ANis Laboratory Turbidimeters. USEPA accepted for calibration purposes. Stablcal Stabilized Formazin Turbidity Standards are true Formazin dilutions developed for use in any turbidimeter. Kit includes 100 mL bottles of 0.1, 20, 200, 1000, 4000 NTU standard and a sealed vial of 7500 NTU standard.