Learn More
Hach Company Stablcal Turbidity Standards Calibration Kit, 2100N / N IS Turbidimeter, 500 mL bottles
Catalog No. 501996499 Shop All Hach Company Products
Encompass
USEPA accepted for calibration purposes. Stablcal Stabilized Formazin Turbidity Standards are true Formazin dilutions developed for use in any turbidimeter. Kit includes 500 mL bottles of 0.1, 20, 200, 1000 and 4000 NTU standards.
Catalog No. 50-199-6499 Supplier Hach Company Supplier No. 2662100
USEPA accepted for calibration purposes. Stablcal Stabilized Formazin Turbidity Standards are true Formazin dilutions developed for use in any turbidimeter. Kit includes 500 mL bottles of 0.1, 20, 200, 1000 and 4000 NTU standards.