Learn More
Hach Company Stablcal Turbidity Standards Calibration Kit, 2100AN / AN IS Turbidimeter, 500 mL bottles
Catalog No. 501996473 Shop All Hach Company Products
Encompass
Stablcal Standards Calibration Kit, for 2100AN and 2100ANis Laboratory Turbidimeters. USEPA accepted for calibration purposes. Stablcal Stabilized Formazin Turbidity Standards are true Formazin dilutions developed for use in any turbidimeter. Kit includes 500 mL bottles of 0.1, 20, 200, 1000, 4000 NTU standard and a sealed vial of 7500 NTU standard.
Catalog No. 50-199-6473 Supplier Hach Company Supplier No. 2659500
Stablcal Standards Calibration Kit, for 2100AN and 2100ANis Laboratory Turbidimeters. USEPA accepted for calibration purposes. Stablcal Stabilized Formazin Turbidity Standards are true Formazin dilutions developed for use in any turbidimeter. Kit includes 500 mL bottles of 0.1, 20, 200, 1000, 4000 NTU standard and a sealed vial of 7500 NTU standard.