Learn More
Hach Company Stablcal Turbidity Standards Calibration Kit,, 2100A Turbidimeter, 100 mL bottles
Catalog No. 501996469 Shop All Hach Company Products
Encompass
Stablcal Standards Calibration Kit, for 2100A Laboratory Turbidimeter. USEPA accepted for calibration purposes. Stablcal Stabilized Formazin Turbidity Standards are true Formazin dilutions developed for use in any turbidimeter. Kit includes 100 mL bottles of 1.0, 10, 100 and 1000 NTU standard.
Catalog No. 50-199-6469 Supplier Hach Company Supplier No. 2659110
Stablcal Standards Calibration Kit, for 2100A Laboratory Turbidimeter. USEPA accepted for calibration purposes. Stablcal Stabilized Formazin Turbidity Standards are true Formazin dilutions developed for use in any turbidimeter. Kit includes 100 mL bottles of 1.0, 10, 100 and 1000 NTU standard.