Learn More
Hach Company Stablcal calibration set with RFID
Catalog No. NC1468089 Shop All Hach Company Products
Encompass
Stablcal calibration set with RFID, for TU5200, TU5300sc, and TU5400sc Laser Turbidimeter. Precise, Pre-mixed Formazin Solutions. Based on Formazin Primary Standards. Enhanced Stability Saves Time and Labor. Confidence in results.
Catalog No. NC1468089 Supplier Hach Company Supplier No. LZY835
Stablcal calibration set with RFID, for TU5200, TU5300sc, and TU5400sc Laser Turbidimeter. Precise, Pre-mixed Formazin Solutions. Based on Formazin Primary Standards. Enhanced Stability Saves Time and Labor. Confidence in results.