Learn More
Hach Company Stablcal Turbidity Standard, 0.50 NTU, 100 mL
Catalog No. 501996569 Shop All Hach Company Products
Encompass
Stablcal Formazin Turbidity Standard, 0.50 NTU. 100 mL Bottle. For TL23, TU52, 2100Q, 2100N, 2100AN series.
Catalog No. 50-199-6569 Supplier Hach Company Supplier No. 2698042
Stablcal Formazin Turbidity Standard, 0.50 NTU. 100 mL Bottle. For TL23, TU52, 2100Q, 2100N, 2100AN series.