Learn More
Hach Company Hach Stablcal Primary Standards Set with RFID, 1 NTU / 10 NTU / 20 NTU (for TU5-series turbidimeters)
Catalog No. NC2830090 Shop All Hach Company Products
Encompass
Stablcal Primary Standards Set with RFID provides stabilized, pre-mixed Formazin primary standards for accurate and repeatable turbidity calibrations. The sealed vial set includes RFID tags for easy identification and reduces time and labor compared with lab-prepared standards.
  • Provides precise, pre-mixed Formazin solutions for calibration.
  • Based on Formazin primary standards to ensure measurement equivalence.
  • Enhanced stability for repeatable results and reduced laboratory preparation.
  • Includes three sealed standards with RFID tags for quick identification.
  • Offers a 400-day shelf life for extended usability.
Catalog No. NC2830090 Supplier Hach Company Supplier No. LZZ005
Stablcal Primary Standards Set with RFID provides stabilized, pre-mixed Formazin primary standards for accurate and repeatable turbidity calibrations. The sealed vial set includes RFID tags for easy identification and reduces time and labor compared with lab-prepared standards.
  • Provides precise, pre-mixed Formazin solutions for calibration.
  • Based on Formazin primary standards to ensure measurement equivalence.
  • Enhanced stability for repeatable results and reduced laboratory preparation.
  • Includes three sealed standards with RFID tags for quick identification.
  • Offers a 400-day shelf life for extended usability.