Learn More
Hach Company Stablcal Formazine Reference Suspension I, 3 NTU, 100mL
Catalog No. 501996773 Shop All Hach Company Products
Encompass
Stabilised Formazine reference suspension for the determination of lucidity/opalescence according to European Pharmacopeia 6.0.
Catalog No. 50-199-6773 Supplier Hach Company Supplier No. 2896742
Stabilised Formazine reference suspension for the determination of lucidity/opalescence according to European Pharmacopeia 6.0.