Learn More
Hach Company Stablcal Reference Suspension Set, bottles
Supplier:  Hach Company 2897200
Encompass
Stabilised Formazine reference suspensions for the determination of lucidity/opalescence according to European Pharmacopeia 6.0. Kit includes 100 mL of each: 0.1, 3, 6, 18, 30.
Catalog No. 50-199-6774