Learn More
Hach Company Stablcal Turbidity Standard, ˂0.1 NTU, 1000 mL (1 L)
Catalog No. 501996478 Shop All Hach Company Products
Encompass
Stablcal Turbidity Standard for all Turbidimeters. USEPA accepted for calibration purposes. Stablcal Stabilized Formazin Turbidity Standards are true Formazin dilutions developed for use in any turbidimeter. NTU Value: 0.1. Bottle: 1000 mL.
Catalog No. 50-199-6478 Supplier Hach Company Supplier No. 2659753
Stablcal Turbidity Standard for all Turbidimeters. USEPA accepted for calibration purposes. Stablcal Stabilized Formazin Turbidity Standards are true Formazin dilutions developed for use in any turbidimeter. NTU Value: 0.1. Bottle: 1000 mL.