Learn More
Hach Company Stablcal Turbidity Standard, ˂ 0.1 NTU, 500mL
Catalog No. 501996477 Shop All Hach Company Products
Encompass
Stablcal Turbidity Standard for all Turbidimeters. USEPA accepted for calibration purposes. Stablcal Stabilized Formazin Turbidity Standards are true Formazin dilutions developed for use in any turbidimeter. NTU Value: 0.1. Bottle: 500ml. For TL23, TU52, 2100Q, 2100N, 2100AN series.
Catalog No. 50-199-6477 Supplier Hach Company Supplier No. 2659749
Stablcal Turbidity Standard for all Turbidimeters. USEPA accepted for calibration purposes. Stablcal Stabilized Formazin Turbidity Standards are true Formazin dilutions developed for use in any turbidimeter. NTU Value: 0.1. Bottle: 500ml. For TL23, TU52, 2100Q, 2100N, 2100AN series.