Learn More
Hach Company Stablcal Turbidity Standard, 20 NTU, 100 mL
Catalog No. 501996486 Shop All Hach Company Products
Encompass
Stablcal Turbidity Standard for all Turbidimeters. USEPA accepted for calibration purposes. Stablcal Stabilized Formazin Turbidity Standards are true Formazin dilutions developed for use in any turbidimeter. NTU Value: 20. Bottle: 100 mL. For TL23, TU52, 2100Q, 2100N, 2100AN series.
Catalog No. 50-199-6486 Supplier Hach Company Supplier No. 2660142
Stablcal Turbidity Standard for all Turbidimeters. USEPA accepted for calibration purposes. Stablcal Stabilized Formazin Turbidity Standards are true Formazin dilutions developed for use in any turbidimeter. NTU Value: 20. Bottle: 100 mL. For TL23, TU52, 2100Q, 2100N, 2100AN series.