Learn More
Hach Company Stablcal, Turbidity Standard, 40.0 NTU, Bottle/1000 mL
Catalog No. 501996614 Shop All Hach Company Products
Encompass
Stablcal Turbidity Standard for all Turbidimeters. Stablcal Stabilized Formazin Turbidity Standards are true Formazin dilutions developed for use in any turbidimeter. NTU Value: 40.0. Bottle: 1000 mL (1 L).
Catalog No. 50-199-6614 Supplier Hach Company Supplier No. 2746353
Stablcal Turbidity Standard for all Turbidimeters. Stablcal Stabilized Formazin Turbidity Standards are true Formazin dilutions developed for use in any turbidimeter. NTU Value: 40.0. Bottle: 1000 mL (1 L).