Learn More
Hach Company Stablcal Turbidity Standard, 40 NTU, 3.78 L
Catalog No. 501996615 Shop All Hach Company Products
Encompass
Stablcal Turbidity Standard for all Turbidimeters. Stablcal Stabilized Formazin Turbidity Standards are true Formazin dilutions developed for use in any turbidimeter. NTU Value: 40.0. Bottle: 3.78 L.
Catalog No. 50-199-6615 Supplier Hach Company Supplier No. 2746356
Stablcal Turbidity Standard for all Turbidimeters. Stablcal Stabilized Formazin Turbidity Standards are true Formazin dilutions developed for use in any turbidimeter. NTU Value: 40.0. Bottle: 3.78 L.