Learn More
Hach Company Stablcal Turb Calibration Kit
Supplier:  Hach Company 2659600
Encompass
Stablcal Turbidity Standard Bulk Solution Calibration Kits for Hach 1720 Series Turbidimeters USEPA accepted for calibration purposes Stablcal Stabilized Formazin Turbidity Standards are true Formazin dilutions developed for use in any turbidimeter Stablcal Kit includes 4 each 1 liter bottles of 0 1 NTU and 20 NTU
Catalog No. 50-002-56944