Learn More
Hach Company Stablcal Turbidity Standard
Supplier:  Hach Company 2660453
Encompass
Ready to use stabilized Formazin Primary standard solution in 1000 mL bottle 200 NTU For TL23 TU52 2100Q 2100N 2100AN series
Catalog No. 50-002-56952