Learn More
Hach Company Stablcal Turbidity Standard
Catalog No. 5000256962 Shop All Hach Company Products
Encompass
Ready to use stabilized Formazin Primary standard solution in 1000 mL bottle 1000 NTU For TL23 TU52 2100Q 2100N 2100AN series
Catalog No. 50-002-56962 Supplier Hach Company Supplier No. 2660653
Ready to use stabilized Formazin Primary standard solution in 1000 mL bottle 1000 NTU For TL23 TU52 2100Q 2100N 2100AN series