Learn More
Hach Company Stablcal Turbidity Standard
Catalog No. 5000256965 Shop All Hach Company Products
Encompass
Ready to use stabilized Formazin Primary standard solution in 1000 mL bottle 800 NTU For TL23 TU52 2100Q 2100N 2100AN series
Catalog No. 50-002-56965 Supplier Hach Company Supplier No. 2660553
Ready to use stabilized Formazin Primary standard solution in 1000 mL bottle 800 NTU For TL23 TU52 2100Q 2100N 2100AN series