Learn More
Hach Company Stablcal Verification 1 NTU
Supplier:  Hach Company LZZ004
Encompass
Stablcal Verification Vial-1 NTU-without RFIDWith proprietary manufacturing technology Hach prepares Stablcal Standards in precise concentrations at a wide variety of concentrations - providing accurate turbidimeter calibrations These standards are compatible with the TU5200 TU5300sc and TU5400sc turbidimeter models ensuring seamless integration and optimal performance The compatibility with these models allows for precise and reliable calibration enhancing the accuracy and efficiency of your turbidity measurements
Catalog No. 50-002-06993