Learn More
Hach Company Stablcal Verification 1 NTU
Supplier:  Hach Company LZZ003
Encompass
Stablcal Verification Vial-1 NTU-with RFID-TU5200With proprietary manufacturing technology Hach prepares Stablcal Standards in precise concentrations at a wide variety of concentrations - providing accurate turbidimeter calibrations
Catalog No. 50-002-07119