Learn More
Hach Company Stablcal Verification 10 NTU
Supplier:  Hach Company LZY877
Encompass
Stablcal Verification Vial-10 NTU-with RFID for TU5200-TU5300sc-and TU5400sc Laser TurbidimetersNew and Improved 400 day Shelf Life
Catalog No. 50-002-07804