Learn More
Hach Company Stablcal Verification 20 NTU
Catalog No. 5000206990 Shop All Hach Company Products
Encompass
Stablcal Calibration Vial-20 NTU-without RFID for TU5200-TU5300sc-and TU5400sc Laser TurbidimetersNew and Improved 400 day Shelf Life
Catalog No. 50-002-06990 Supplier Hach Company Supplier No. LZY899
Stablcal Calibration Vial-20 NTU-without RFID for TU5200-TU5300sc-and TU5400sc Laser TurbidimetersNew and Improved 400 day Shelf Life