Learn More
Hach Company Stablcal Verification 600 NTU
Catalog No. 5000207120 Shop All Hach Company Products
Encompass
Stablcal Calibration Vial-600 NTU-without RFID for TU5200-TU5300sc-and TU5400sc Laser TurbidimetersNew and Improved 400 day Shelf Life
Catalog No. 50-002-07120 Supplier Hach Company Supplier No. LZY900
Stablcal Calibration Vial-600 NTU-without RFID for TU5200-TU5300sc-and TU5400sc Laser TurbidimetersNew and Improved 400 day Shelf Life