Learn More
Diagnostic Biosystems STABLE DAB/PLUS Stable DAB/Plus
Catalog No. NC2049142
Encompass_Preferred
DAB, a widely used chromogen for immunoperoxidase staining, is well accepted among pathologists because of its increased sensitivity and ability to give cleaner background as compared to amino ethylcarbazole (AEC). Specimens stained in DAB can be dehydrated, cleared, and mounted for permanent record keeping. Stable DAB/Plus is more sensitive and stable than traditional working DAB solutions.
Catalog No. NC2049142 Supplier Diagnostic Biosystems Supplier No. K047
DAB, a widely used chromogen for immunoperoxidase staining, is well accepted among pathologists because of its increased sensitivity and ability to give cleaner background as compared to amino ethylcarbazole (AEC). Specimens stained in DAB can be dehydrated, cleared, and mounted for permanent record keeping. Stable DAB/Plus is more sensitive and stable than traditional working DAB solutions.