Learn More
Bachem ([13C6]Leu5)-Ghrelin (human) Trifluoroacetate
SDP
Supplier:  Bachem 4072025.0500
Encompass_Preferred
([13C6]Leu5)-Ghrelin (human) Trifluoroacetate, H-Gly-Ser-Ser(octanoyl)-Phe-[13C6]Leu-Ser-Pro-Glu-His-Gln-Arg-Val-Gln-Gln-Arg-Lys-Glu-Ser-Lys-Lys-Pro-Pro-Ala-Lys-Leu-Gln-Pro-Arg-OH, 1802078-40-5, C14313C6H249N47O42, Mr 3376.84, 0.5mg. Stable isotope-labeled human ghrelin useful for pharmacokinetic/pharmacodynamic studies.
Catalog No. 50-194-6891