Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poltex Stable Stack Tip Box Shelf
SDP

Catalog No. 01259155
Change view
Click to view available options
Mounting Type:
Magnet Mount
Tape Mount
Wall Mount
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Mounting Type
01-259-155 Magnet Mount
01-259-141 Tape Mount
01-259-217 Wall Mount
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
N/A
Catalog No. 01-259-155 Supplier Poltex Supplier No. STASTAKSHFM
Add to Cart
Add to Cart
N/A

Shelf for stable stack pipette tips. Choose from Wall Mount, HB Tape Mount, or Magnet Mount with durable and cleanable PETG construction.

  • Shelf for stable stack pipette tips.
  • Choose from Wall Mount (screws/anchors included), HB Tape Mount (high bond tape included), or Magnet Mount (powerful magnets included).
  • Durable and cleanable PETG construction.
  • Consumables not included.

Specifications

For Use With (Application) Shelf for Stable Stack Pipette Tips
Quantity 1 Ea.
Color Clear
Dimensions (L x W x H) 5.4 x 14.4 x 11.45 in.
Autoclavable Not Autoclavable
Mounting Type Magnet Mount
Holds Pipette Tips
Material PETG
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.