Learn More
Agilent Technologies STABLEBND 300 GRD CRTRDG, 4/PK
Catalog No. NC2161589
Encompass
Poroshell StableBond 300 C3, 2.1 x 12.5 mm, 5 µm, guard cartridge (ZGC) 4/pk. Use with hardware kit 820999-901.
This item is not returnable. View return policy
Catalog No. NC2161589 Supplier Agilent Technologies Supplier No. 821075924
Poroshell StableBond 300 C3, 2.1 x 12.5 mm, 5 µm, guard cartridge (ZGC) 4/pk. Use with hardware kit 820999-901.