Learn More
Sigma Aldrich Fine Chemicals Biosciences StableCell(TM) DMEM - high glucose With 4500 mg/L glucose, stable glutamine, and sodium bicarbonate, without sodium pyruvate., liquid, sterile-filtered, suitable for cell culture | 500ML
Catalog No. NC1979439
Encompass
StableCell(TM) DMEM - high glucose With 4500 mg/L glucose, stable glutamine, and sodium bicarbonate, without sodium pyruvate., liquid, sterile-filtered, suitable for cell culture | 500ML
Catalog No. NC1979439 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. D0819500ML
StableCell(TM) DMEM - high glucose With 4500 mg/L glucose, stable glutamine, and sodium bicarbonate, without sodium pyruvate., liquid, sterile-filtered, suitable for cell culture | 500ML