Learn More
Sigma Aldrich Fine Chemicals Biosciences StableCellTM DMEM/f12 Wth500ML
Catalog No. 502901032
Encompass
This DMEM f-12 Ham medium differs from the original formula because it is supplemented with L-alanyl-L-glutamine dipeptide. Glutamine is notoriously unstable in cell culture. This dipeptide provides a more stable form of glutamine for use in cell cultures.
Catalog No. 50-290-1032 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. D0697500ML
This DMEM f-12 Ham medium differs from the original formula because it is supplemented with L-alanyl-L-glutamine dipeptide. Glutamine is notoriously unstable in cell culture. This dipeptide provides a more stable form of glutamine for use in cell cultures.