Learn More
Sigma Aldrich Fine Chemicals Biosciences StableCellTM DMEM - hgh g500ML
Catalog No. 502901360
Encompass
This DMEM-Hi glucose medium differs from the original formula because it is supplemented with L-alanyl-L-glutamine dipeptide. Glutamine is notoriously unstable in cell culture. This dipeptide provides a more stable form of glutamine for use in cell cultures.
Catalog No. 50-290-1360 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. D0822500ML
This DMEM-Hi glucose medium differs from the original formula because it is supplemented with L-alanyl-L-glutamine dipeptide. Glutamine is notoriously unstable in cell culture. This dipeptide provides a more stable form of glutamine for use in cell cultures.