Learn More
Cole-Parmer Cole-Parmer StableTemp Digital General-Purpose Water Bath, 5 L; 120 VAC
Catalog No. NC1630902 Shop All Cole-Parmer Products
Encompass
Cole-Parmer StableTemp utility baths combine precise temperature control and excellent temperature stability to provide you with reliable heating. These baths feature dual thermostats — a primary microprocessor-based temperature controller that provides ±0.1°C stability (±0.2°C uniformity), and a secondary over-temperature safety sensor prevents samples from overheating. For optimal temperature accuracy, the primary controller can be calibrated to your reference thermometer. The stainless steel tank is seamless—no welds to leak or rust—and the rounded corners provide an easy-to-clean interior. The high-dome, hinged polycarbonate lid tilts out of the way, while allowing complete view of the sample and condensation to drain back. Select operation in English, Spanish, French, Chinese, or German. 10-, 20-, and 28-liter models also feature rear-mounted drain.
Catalog No. NC1630902 Supplier Cole-Parmer Supplier No. 1210586
Cole-Parmer StableTemp utility baths combine precise temperature control and excellent temperature stability to provide you with reliable heating. These baths feature dual thermostats — a primary microprocessor-based temperature controller that provides ±0.1°C stability (±0.2°C uniformity), and a secondary over-temperature safety sensor prevents samples from overheating. For optimal temperature accuracy, the primary controller can be calibrated to your reference thermometer. The stainless steel tank is seamless—no welds to leak or rust—and the rounded corners provide an easy-to-clean interior. The high-dome, hinged polycarbonate lid tilts out of the way, while allowing complete view of the sample and condensation to drain back. Select operation in English, Spanish, French, Chinese, or German. 10-, 20-, and 28-liter models also feature rear-mounted drain.