Learn More
Aviva Systems Biology Corp STAC Antibody - middle region (ARP64320_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64320_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against STAC. It was validated on Western Blot. . Alternate names: FLJ32331, STAC1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-288-234