Learn More
Enzo Life Sciences Stachybotrylactam (0.5mg). CAS: 163391-76-2
SDP
Supplier:  Enzo Life Sciences ALX630112MC05
Encompass_Preferred
Mycotoxin. Immunosuppressive. Weakly inhibits HIV-1 protease. Alternative name: 2-Deoxy Antibiotic F1839A. Purity: ≥95% (HPLC). Solubility: Soluble in 100% ethanol, methanol, dimethyl formamide or DMSO. Long Term Storage: -20°C.
Catalog No. 50-200-9637