Learn More
Cayman Chemical Stachybotrylactam 2.5mg
SDP
Catalog No. 502987730
Encompass_Preferred
A mycotoxin with antiviral activity; it inhibits HIV-1 protease in vitro (IC50 = 161 μM); no observable cytotoxicity at concentrations up to 100 μM in HepG2 cells
Catalog No. 50-298-7730 Supplier Cayman Chemical Supplier No. 225842.5 MG
A mycotoxin with antiviral activity; it inhibits HIV-1 protease in vitro (IC50 = 161 μM); no observable cytotoxicity at concentrations up to 100 μM in HepG2 cells