Learn More
Medchemexpress LLC Stachydrine hydrochloride | 4136-37-2 | 179.64 | 500 MG
SDP
Catalog No. 5000416209
Encompass_Preferred
Stachydrine hydrochloride is the major active constituent of *Leonurus artemisia*, a potential therapy for cardiovascular diseases. It can inhibit the NF-κB signal pathway, exhibiting anti-hypertrophic activities.
  • Major active constituent of *Leonurus artemisia*
  • Potential therapy for cardiovascular diseases
  • Inhibits the NF-κB signal pathway
  • Exhibits anti-hypertrophic activities
  • Suppresses p-IκB (ser32) protein in cytosol
  • Suppresses NF-κB (p65) protein in nucleus
  • Inhibits MCF-7 and T47D cell proliferation in a dose- and time-dependent manner
  • Increases frequency of MCF-7 and T47D cells at G1 phase of cell cycle, suggesting cell cycle arrest
Catalog No. 50-004-16209 Supplier Medchemexpress LLC Supplier No. HYN0738500MG
Stachydrine hydrochloride is the major active constituent of *Leonurus artemisia*, a potential therapy for cardiovascular diseases. It can inhibit the NF-κB signal pathway, exhibiting anti-hypertrophic activities.
  • Major active constituent of *Leonurus artemisia*
  • Potential therapy for cardiovascular diseases
  • Inhibits the NF-κB signal pathway
  • Exhibits anti-hypertrophic activities
  • Suppresses p-IκB (ser32) protein in cytosol
  • Suppresses NF-κB (p65) protein in nucleus
  • Inhibits MCF-7 and T47D cell proliferation in a dose- and time-dependent manner
  • Increases frequency of MCF-7 and T47D cells at G1 phase of cell cycle, suggesting cell cycle arrest