Learn More
Medchemexpress LLC Stachydrine hydrochloride | 4136-37-2 | MFCD00060174 | 179.64 | 1 ML
SDP
Catalog No. 5000417147
Encompass_Preferred
Stachydrine hydrochloride, a major active constituent of Leonurus artemisia, is a potential therapy for cardiovascular diseases. It inhibits the NF-κB signal pathway and demonstrates anti-hypertrophic activities. Research indicates it can suppress p-IκB (ser32) protein and NF-κB (p65) protein, and inhibit MCF-7 and T47D cell proliferation.
  • Inhibits NF-κB signal pathway
  • Exhibits anti-hypertrophic activities
  • Suppresses p-IκB (ser32) protein in cytosol
  • Suppresses NF-κB (p65) protein in nucleus
  • Inhibits MCF-7 and T47D cell proliferation
  • Induces G1 phase cell cycle arrest in cell lines
Catalog No. 50-004-17147 Supplier Medchemexpress LLC Supplier No. HYN073810MM/1ML
Stachydrine hydrochloride, a major active constituent of Leonurus artemisia, is a potential therapy for cardiovascular diseases. It inhibits the NF-κB signal pathway and demonstrates anti-hypertrophic activities. Research indicates it can suppress p-IκB (ser32) protein and NF-κB (p65) protein, and inhibit MCF-7 and T47D cell proliferation.
  • Inhibits NF-κB signal pathway
  • Exhibits anti-hypertrophic activities
  • Suppresses p-IκB (ser32) protein in cytosol
  • Suppresses NF-κB (p65) protein in nucleus
  • Inhibits MCF-7 and T47D cell proliferation
  • Induces G1 phase cell cycle arrest in cell lines